Learn More
Rice Lake Weighing System WEIGHT KIT 200G ADJUSTED
Catalog No. 50809407
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems, 200g Adjusted, ASTM Class 1, SST Polished, Density 7.84, Electronic Balance Calibration Weights
Catalog No. 50-809-407 Supplier Rice Lake Weighing System Supplier No. 12948
Calibration Weight Kit, Rice Lake Weighing Systems, 200g Adjusted, ASTM Class 1, SST Polished, Density 7.84, Electronic Balance Calibration Weights