Learn More
Rice Lake Weighing System Weight Set,2kg-1g ASTM Class 5
Catalog No. 50809353
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems, 2kg-1g, ASTM Class 5, Stainless Steel, Satin finish, (5-2-2-1 configuration), 4.11kg Kit
Catalog No. 50-809-353 Supplier Rice Lake Weighing System Supplier No. 12524
Calibration Weight Kit, Rice Lake Weighing Systems, 2kg-1g, ASTM Class 5, Stainless Steel, Satin finish, (5-2-2-1 configuration), 4.11kg Kit