Learn More
Rice Lake Weighing System WEIGHT KIT 2MG ASTM CLASS 0
Catalog No. 50809433
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems, 2mg, ASTM Class 0, Density 2.7
Catalog No. 50-809-433 Supplier Rice Lake Weighing System Supplier No. 45884
Calibration Weight Kit, Rice Lake Weighing Systems, 2mg, ASTM Class 0, Density 2.7