Learn More
Rice Lake Weighing System Weight Set,50g-1mg ASTM Class 5
Catalog No. 50809341
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems, 50g-1mg, ASTM Class 5, Stainless Steel, Satin finish, (5-2-2-1 configuration), 111.11g Kit
Catalog No. 50-809-341 Supplier Rice Lake Weighing System Supplier No. 12504
Calibration Weight Kit, Rice Lake Weighing Systems, 50g-1mg, ASTM Class 5, Stainless Steel, Satin finish, (5-2-2-1 configuration), 111.11g Kit