Learn More
Rice Lake Weighing System WEIGHT KIT 50G ADJUSTED ASTM
Catalog No. 50809411
Encompass_Preferred
Calibration Weight Kit, Rice Lake Weighing Systems, 50g, Adjusted, ASTM Class 1, SST Polished, Density 7.84, Electronic Balance Calibration Weights
Catalog No. 50-809-411 Supplier Rice Lake Weighing System Supplier No. 12964
Calibration Weight Kit, Rice Lake Weighing Systems, 50g, Adjusted, ASTM Class 1, SST Polished, Density 7.84, Electronic Balance Calibration Weights