Learn More
Rice Lake Weighing System Weight, 2mg ASTM Class 5
Catalog No. 50809404
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Leaf Style, 2mg, ASTM Class 5, Stainless Steel, Satin finish, Density 7.95
Catalog No. 50-809-404 Supplier Rice Lake Weighing System Supplier No. 12923
Calibration Weight, Rice Lake Weighing Systems, Leaf Style, 2mg, ASTM Class 5, Stainless Steel, Satin finish, Density 7.95