Learn More
Rice Lake Weighing System Weight, 500mg ASTM Class 5
Catalog No. 50809398
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Leaf Style, 500mg, ASTM Class 5, Stainless Steel, Satin finish, Density 7.95
Catalog No. 50-809-398 Supplier Rice Lake Weighing System Supplier No. 12913
Calibration Weight, Rice Lake Weighing Systems, Leaf Style, 500mg, ASTM Class 5, Stainless Steel, Satin finish, Density 7.95