Learn More
Rice Lake Weighing System WEIGHT SCREW KNOB 100G ASTM
Catalog No. 50809332
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Screw Knob, 100g, ASTM, Class 1, SST, Polished Density 7.95
Catalog No. 50-809-332 Supplier Rice Lake Weighing System Supplier No. 11864
Calibration Weight, Rice Lake Weighing Systems, Screw Knob, 100g, ASTM, Class 1, SST, Polished Density 7.95