Learn More
Rice Lake Weighing System WEIGHT SCREW KNOB 10G ASTM
Catalog No. 50809329
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Screw Knob, 10g, ASTM, Class 1, SST, Polished Density 7.95
Catalog No. 50-809-329 Supplier Rice Lake Weighing System Supplier No. 11859
Calibration Weight, Rice Lake Weighing Systems, Screw Knob, 10g, ASTM, Class 1, SST, Polished Density 7.95