Learn More
Rice Lake Weighing System WEIGHT SCREW KNOB 5G ASTM
Catalog No. 50809328
Encompass_Preferred
Calibration Weight, Rice Lake Weighing Systems, Screw Knob, 5g, ASTM, Class 1, SST, Polished Density 7.95
Catalog No. 50-809-328 Supplier Rice Lake Weighing System Supplier No. 11857
Calibration Weight, Rice Lake Weighing Systems, Screw Knob, 5g, ASTM, Class 1, SST, Polished Density 7.95