Learn More
Hach Company Weight Set, Class M-2
Supplier:  Hach Company 2101100
Encompass
Standard weight set. 16-piece, with stainless steel forceps. Eight fractional weights: 10, 20, 50, 100, 200, 200, 200, and 500 mg. (Two fractional weights, 200 mg and 500mg, are stainless steel, the rest are aluminum) Eight brass gram weighs: 1, 2, 2, 5, 10, 10, 20, and 50 grams. Polypropylene case.
Catalog No. 50-199-5384