Learn More
Scientific Industries WEIGHT SET 50G-10MG
Catalog No. 50728294
Encompass_Preferred
50g; 10mg Metric & Apothecary Weight Set. Stainless steel set furnished in hinged, cover box with stainless steel forceps. This weight set is adjusted to N.B.S. class C Tolerances.
This item is not returnable. View return policy
Catalog No. 50-728-294 Supplier Scientific Industries Supplier No. TC105
50g; 10mg Metric & Apothecary Weight Set. Stainless steel set furnished in hinged, cover box with stainless steel forceps. This weight set is adjusted to N.B.S. class C Tolerances.