Learn More
GSC Go Science Crazy Weight Set, Hooked 9 pieces
SCI_ED
Catalog No. S24634 Shop All GSC Products
Click to view available options
Quantity:
9 Pieces
Catalog No. Quantity
S24634 9 Pieces
1 options
Catalog No. S24634 Supplier GSC Go Science Crazy Supplier No. 425001
Quality accurately calibrated weight set with case
Nine Piece Hooked Weight Set with two cast iron hooked weights (1000g, 500g), seven brass hooked weights (two 200g, one 100g, one 50g, two20g, one 10g), and case
Specifications
Material Brass and Iron
Quantity 9 Pieces
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov