Learn More
Electron Microscopy Sciences Weighted Beaker/Flask Holder, 100 ml, 165 g Weight, 60 mm diameter
SDP
Catalog No. 501899172
Encompass_Preferred
This weighted holder grips the beaker or flask firmly in four flared stainless steel prongs. The Vikem vinyl-coated lead ring secured beneath the beaker keeps it immersed in water, providing a low center of gravity which allows water to circulate underneath the beaker.
Use: 100 ml
Weight: 165 g
Diameter: 60 mm
Catalog No. 50-189-9172 Supplier Electron Microscopy Sciences Supplier No. 6444101
This weighted holder grips the beaker or flask firmly in four flared stainless steel prongs. The Vikem vinyl-coated lead ring secured beneath the beaker keeps it immersed in water, providing a low center of gravity which allows water to circulate underneath the beaker.
Use: 100 ml
Weight: 165 g
Diameter: 60 mm