Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Weighted Equivalency Sets

Catalog No. S90839B
Change view
Click to view available options
Decimals Set
Fractions Set
Percentages Set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S90839B Decimals Set
S90839A Fractions Set
S90839C Percentages Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S90839B Supplier Learning Resources, Inc Supplier No. LER3021

Includes

  • 24 number pieces
  • Activity guides

Perfect for use with pan balances in class demonstration or small-group activities.

  • Make the relationship between fractions, decimals, and percents easy to understand
  • Colorful, plastic numbers are weighted, relative to the whole (100%, 50%, 25%, etc.)
  • Sets are interchangeable
  • Color coding aligns with the Rainbow Fraction™ System
  • Each number measures approximately 3 x 1 1/2 in.

Order Info

Grades K-5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.