Learn More
Cole-Parmer Scienceware F24962-0000 Weighted safety shield, 30" height
Catalog No. NC0306388 Shop All Cole-Parmer Products
Encompass_Preferred
Shields are constructed of 3/16" thick, clear polycarbonate to protect from splash and flying debris. Three-sided construction with epoxy-coated, steel base provides added stability and coverage.
Catalog No. NC0306388 Supplier Cole-Parmer Supplier No. 8646910
Shields are constructed of 3/16" thick, clear polycarbonate to protect from splash and flying debris. Three-sided construction with epoxy-coated, steel base provides added stability and coverage.