Learn More
Cole-Parmer Anti-static polypropylene weighing funnel, micro
Catalog No. NC9751277 Shop All Cole-Parmer Products
Encompass_Preferred
Ensure complete sample transfer with these disposable polypropylene (PP) weighing funnels that are designed to fit in volumetric flasks. One side of the funnel is flattened so you can set the funnel flat on a balance. Micro funnel is designed to fit microwell plates. Autoclavable.
Catalog No. NC9751277 Supplier Cole-Parmer Supplier No. 0612700
Ensure complete sample transfer with these disposable polypropylene (PP) weighing funnels that are designed to fit in volumetric flasks. One side of the funnel is flattened so you can set the funnel flat on a balance. Micro funnel is designed to fit microwell plates. Autoclavable.