Learn More
Mettler Toledo™ WeightLink™ DMC Scanner
Catalog No. 01914369 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Scanner; Mettler Toledo; WeightLink DMC Scanner
Catalog No.
01-914-369 Scanner; Mettler Toledo; WeightLink DMC Scanner
1 options
Catalog No. 01-914-369 Supplier Mettler Toledo™ Supplier No. 30268560
Image analysis technology is used to scan smallest data matrix codes on reflective surfaces such as Mettler Toledo WeightLink™ Weights. Hands-Free Operation - The integrated optical sensors enables a contactless operation of the device. Made of aluminum and the polycarbonate cover makes it suitable for all kinds of environments.
  • Interfaces: RS232
  • Material: Polycarbonate;Aluminum
  • Technology: Image Analysis
  • Best Scanning Performance - Image analysis technology is used to scan smallest data matrix codes on reflective surfaces such as Mettler Toledo WeightLink™ Weights.
  • Hands-Free Operation - The integrated optical sensors enables a contactless operation of the device.
  • Robust Design - Made of aluminum and the polycarbonate cover makes it suitable for all kinds of environments.
TRUSTED_SUSTAINABILITY
Specifications
Product Type DMC Scanner