Learn More
MSC Weiler Scrub Brush, 6 in, Finger Grip Handle, Polypropylene, Medium Firmness
Catalog No. 502839215
Encompass
This scrub brush features a 6 inch length with a finger grip handle and white polypropylene fill. It is ideal for heavy-duty scrubbing applications.
  • Ideal for heavy-duty scrubbing.
  • Features an iron style plastic block.
  • Constructed with polypropylene fill material.
  • Provides medium fill firmness.
  • Equipped with white fill color.
  • Has a trim length of 1-1/8 in.
  • Includes a plastic block material.
  • Handle length of 6 in.
Catalog No. 50-283-9215 Supplier MSC Supplier No. 82071226
This scrub brush features a 6 inch length with a finger grip handle and white polypropylene fill. It is ideal for heavy-duty scrubbing applications.
  • Ideal for heavy-duty scrubbing.
  • Features an iron style plastic block.
  • Constructed with polypropylene fill material.
  • Provides medium fill firmness.
  • Equipped with white fill color.
  • Has a trim length of 1-1/8 in.
  • Includes a plastic block material.
  • Handle length of 6 in.