Learn More
MSC Weiler Pencil End Brush, 1/4 in, Steel Fill, 1 ea
Catalog No. 502870178
Encompass
This 1/4 in Pencil End Brush features straight wire fill material retained within a small diameter steel tube. It provides a light-duty brushing action, making it suitable for use in very small recessed areas, and is designed for use with drills and drill presses.
  • Contains straight wire fill material retained in a small diameter steel tube.
  • Provides a light-duty brushing action suitable for very small recessed areas.
  • Suitable for cleaning, deburring, and finishing applications.
  • Compatible with corded/cordless hand drills, drill presses, drilling machines, right-angle grinders, and CNC/milling/automatic machines.
Catalog No. 50-287-0178 Supplier MSC Supplier No. 82074691
This 1/4 in Pencil End Brush features straight wire fill material retained within a small diameter steel tube. It provides a light-duty brushing action, making it suitable for use in very small recessed areas, and is designed for use with drills and drill presses.
  • Contains straight wire fill material retained in a small diameter steel tube.
  • Provides a light-duty brushing action suitable for very small recessed areas.
  • Suitable for cleaning, deburring, and finishing applications.
  • Compatible with corded/cordless hand drills, drill presses, drilling machines, right-angle grinders, and CNC/milling/automatic machines.