Learn More
Sklar Instruments Weinstein Instrument Holder, Sklar, 10 in., Straight, Smooth, Stainless Steel. This holder is 2-1/2 in. x 10 in. EA/1.
SDP
Catalog No. 501437166 Shop All Sklar Instruments Products
Encompass_Preferred
Weinstein Instrument Holder, Sklar, 10 in., Straight, Smooth, Stainless Steel. Sklar's Weinstein Instrument Holder is designed to expand instruments for greater expose hard to reach areas during cleaning and sterilization. It is available in a variety of lengths and widths. This holder is 2-1/2 inches x 10 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7166 Supplier Sklar Instruments Supplier No. 102010
Weinstein Instrument Holder, Sklar, 10 in., Straight, Smooth, Stainless Steel. Sklar's Weinstein Instrument Holder is designed to expand instruments for greater expose hard to reach areas during cleaning and sterilization. It is available in a variety of lengths and widths. This holder is 2-1/2 inches x 10 inches.