Learn More
Fine Science Tools Fine Science Tools Weitlaner Retractor, Surgical Stainless Steel, 2x3 Blunt Teeth, 10.5 cm, 1 ea
SDP
Catalog No. NC3917983
Encompass
The Weitlaner retractor is a specialized surgical instrument designed for tissue retraction in non-human research applications. Fabricated from high-quality surgical stainless steel (Inox), this straight-pattern retractor provides reliable performance and durability.
  • Constructed from surgical stainless steel (Inox)
  • Features 2 x 3 blunt teeth
  • Maximum spread capacity of 4 cm
  • Teeth dimensions of 7 mm in length and 3 mm in width
  • Overall length of 10.5 cm
  • Weight of 55 g
  • Designed for research use only, not for human application
  • Manufactured in Germany
Catalog No. NC3917983 Supplier Fine Science Tools Supplier No. 1701211
The Weitlaner retractor is a specialized surgical instrument designed for tissue retraction in non-human research applications. Fabricated from high-quality surgical stainless steel (Inox), this straight-pattern retractor provides reliable performance and durability.
  • Constructed from surgical stainless steel (Inox)
  • Features 2 x 3 blunt teeth
  • Maximum spread capacity of 4 cm
  • Teeth dimensions of 7 mm in length and 3 mm in width
  • Overall length of 10.5 cm
  • Weight of 55 g
  • Designed for research use only, not for human application
  • Manufactured in Germany