Learn More
Med Vet International Miltex Vantage Weitlaner Retractor, 6.5in, With 3X4 Blunt Teeth
Catalog No. 502830412
Encompass_Preferred
The Miltex Vantage V911-616 Weitlaner Retractor has a length of 6.5in. and is made of mid-grade stainless steel. The Weitlaner Retractor is a self-retaining retractor is a useful surgical instrument for small, depp incisions and soft tissue dissection.
Catalog No. 50-283-0412 Supplier Med Vet International Supplier No. MILV911616
The Miltex Vantage V911-616 Weitlaner Retractor has a length of 6.5in. and is made of mid-grade stainless steel. The Weitlaner Retractor is a self-retaining retractor is a useful surgical instrument for small, depp incisions and soft tissue dissection.