Learn More
Stoelting™ Weitlaner Retractor
SDP
Catalog No. 10000491 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
4 in.
4.5 in.
5.5 in.
6.5 in.
8 in.
Model:
Blunt
Sharp prongs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Model
10-000-491 4.5 in. Sharp prongs
10-001-433 4 in. Blunt
10-001-434 5.5 in. Blunt
10-001-435 6.5 in. Blunt
10-001-436 8 in. Blunt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-000-491 Supplier Stoelting™ Supplier No. 5212800P
Only null left
Add to Cart
Add to Cart
Stoelting™ Weitlaner Retractor can be used to actively separate the edges of a surgical incision or wound or hold back underlying organs and tissues so that body parts under the incision may be accessed.
  • Sharp retractors are better at providing a secure grip and they can safely hold back the skin, muscles, fascia, tendons, or the periosteum but they may damage vessels, nerves, the peritoneum, glands or cysts.
  • Blunt retractors are recommended when iatrogenic injury is a concern.
  • Main benefit of blunt retractors is that they prevent unintended injury to the tissue held by the retractor or to the underlying tissue.
Specifications
For Use With (Application) Neuroscience
Length (English) 4.5 in.
Size 2 x 3
Model Sharp prongs