Learn More
WELCH MATERIALS INC Welchrom C18E SPE Cartridges , 200mg/3mL, 50pcs/pk
Catalog No. NC2052803
Encompass_Preferred
SPE Cartridges, Welchrom C18E, 200mg/3mL, 50pcs/pk, brand: Welchrom, bonding phase: C18E, particle size: 40-60 μm, surface area: 480 m2/g, pore size: 60Å, Pore volume: 0.80 cm3/g, carbon loading: 17%, endcapped.
Catalog No. NC2052803 Supplier WELCH MATERIALS INC Supplier No. 0055911004
SPE Cartridges, Welchrom C18E, 200mg/3mL, 50pcs/pk, brand: Welchrom, bonding phase: C18E, particle size: 40-60 μm, surface area: 480 m2/g, pore size: 60Å, Pore volume: 0.80 cm3/g, carbon loading: 17%, endcapped.