Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Adapters
Description
- Bleach resistant construction for extending service life
- Continuous vacuum level monitoring with dial vacuum gauge
- Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
| For Use With | Welch Compact Aspiration/Fitlration Vacuum Station |
| Material | Stainless Steel |
| No. of Channels | 8 |
| Type | Adapter, 8-Channel |