Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Adapters
Catalog No. 01258113
Change view
Click to view available options
Type:
Adapter with 40 mm Tip
Adapter, 8-Channel
Adapter, 8-Channel with Ejector
Adapter, Single Channel
Adapter, for Pasteur Pipets
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-258-113 Adapter, for Pasteur Pipets
01-258-112 Adapter, Single Channel
01-258-110 Adapter with 40 mm Tip
01-258-111 Adapter, 8-Channel
01-258-114 Adapter, 8-Channel with Ejector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-258-113 Supplier Welch™ Supplier No. 1475K04
Add to Cart
Add to Cart
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
For Use With Welch Compact Aspiration/Fitlration Vacuum Station
Material Rubber
Type Adapter, for Pasteur Pipets