Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Adapters
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
$95.40 - $895.50
Specifications
For Use With Welch Compact Aspiration/Fitlration Vacuum Station
View More Specs
Products 5
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
01-258-112
View Documents
Welch™
1475K03
Adapter, Single Channel
Each for $109.90
Only null left
Add to Cart
01-258-110
View Documents
Welch™
1475K01
Adapter with 40 mm Tip
Each for $134.00
Only null left
Add to Cart
01-258-111
View Documents
Welch™
1475K02
Adapter, 8-Channel
Each for $570.00
Only null left
Add to Cart
01-258-113
View Documents
Welch™
1475K04
Adapter, for Pasteur Pipets
Each for $95.40
Only null left
Add to Cart
01-258-114
View Documents
Welch™
1475K05
Adapter, 8-Channel with Ejector
Each for $895.50
Only null left
Add to Cart
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
Welch Compact Aspiration/Fitlration Vacuum Station
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.