Learn More
WELCH MATERIALS INC Welch MaterialsTM Ghost-buster Column, 2.1×50mm
Catalog No. NC1940825
Encompass_Preferred
Welch Ghost-Buster Column can effectively remove impurities with low polarity and thus prevents the interference from all kinds of ghost peaks. It is installed between gradient mixer and injector, which helps remove not only the impurities in mobile phase, but impurities in mixer and pipelines as well. Dimension: 2.1×50mm, suitable for ID 2.1/3.0 mm HPLC column and UHPLC, Max. Pressure: 100 Mpa.
Catalog No. NC1940825 Supplier WELCH MATERIALS INC Supplier No. 0610031025
Welch Ghost-Buster Column can effectively remove impurities with low polarity and thus prevents the interference from all kinds of ghost peaks. It is installed between gradient mixer and injector, which helps remove not only the impurities in mobile phase, but impurities in mixer and pipelines as well. Dimension: 2.1×50mm, suitable for ID 2.1/3.0 mm HPLC column and UHPLC, Max. Pressure: 100 Mpa.