Learn More
Welch™ Hand Held Pipette for Aspiration Stations: Tip Adapters
Catalog No. 01258115
Change view
Click to view available options
Description:
1 channel adapter, With injector, Disposable pipette tip adapter
4-channel adapter, Microtiter format manifolds, 40 mm
Cannula-style needle adapter, 150 mm
Cannula-style needle adapter, 280 mm
For Use With (Equipment):
Aspirator or Filtration Station
Disposable Tip with Injector
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description For Use With (Equipment)
01-258-115 Cannula-style needle adapter, 150 mm Aspirator or Filtration Station
01-258-118 1 channel adapter, With injector, Disposable pipette tip adapter Disposable Tip with Injector
01-258-117 Cannula-style needle adapter, 280 mm Aspirator or Filtration Station
01-258-116 4-channel adapter, Microtiter format manifolds, 40 mm Aspirator or Filtration Station
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-258-115 Supplier Welch™ Supplier No. 1475K06
Only null left
Add to Cart
Add to Cart
Specifications
Description Cannula-style needle adapter, 150 mm
For Use With (Equipment) Aspirator or Filtration Station
Material Stainless Steel