Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Welch™ Hose Adapter Kit for Vacuum Pumps

Catalog No. 0125720
Change view
Click to view available options
For Use With:
Large vacuum ovens
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
01-257-20 Large vacuum ovens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-20 Supplier Welch™ Supplier No. 1420H02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for Welch vaccum pumps

  • Includes: 1.5m (5 ft.) of thick walled rubber vacuum hose
  • For proper sized tubing adapters and vacuum tight connections
  • Pump inlet I.D.: 13/ in.
  • Appliance connector OD: 1/4, 3/8

Specifications

For Use With Large vacuum ovens
Includes 5 ft. Thick walled Rubber vacuum hose, 0.81 in. I.D. inlet, 0.25, 0.37 in. O.D. appliance connector
Type Hose Adapter Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.