Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Welch™ Replacement Exhaust Element for Vacuum Pumps

Catalog No. 011045H
Change view
Click to view available options
Type:
Exhaust Element
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-104-5H Exhaust Element
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-104-5H Supplier Welch™ Supplier No. 1417L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for Welch Vacuum Pump Models 8803, 8804, 8890, 8905, 1399, 1400

For use with Welch Vacuum Pump Models 8803, 8804, 8890, 8905, 1399, 1400

  • Diameter x Height: 6.35 x 6.35cm (2.5 x 2.5 in.)

Specifications

For Use With Model 8803, 8804, 8890, 8905, 1399, 1400 vacuum pump
Overall Height (English) 2.5 in.
Overall Height (Metric) 6.35 cm
Overall Width (English) 2.5 in.
Type Exhaust Element
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.