Learn More
Sigma Aldrich Fine Chemicals Biosciences WelchR ChemStarTM vacuum p1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z4118091EA
Encompass
ChemStar vacuum pumps are distinguished by having all five of the defining features needed for pumping corrosive gases..nbspHigh contamination tolerance large oil reservoir dilutes contaminants.nbspCorrosion resistant parts Viton(R) PTFE stainless steel nickel-plated or anodized parts.nbspReduced frictional wear runs at less than 580 rpm versus 1700 rpm for direct-drive pumps.nbspNitrogen purge standard degasses and cools pump oil reducing corrosion.nbspEasy attachment of accessories trap and oil mist eliminatorSupplied with 2 pin plugModel 1400N specificationsFree Air Displacement 0.9 cfm (25 L/min)Max. vacuum 1 x 10-4 torrOil capacity 0.62 qt (0.59 L)Tubing needed I.D. 7/16 in.Exhaust and intake thread type NW 16Pumping speed 580 rpmDim. L x W x H 17.8 x 9 x 12.6 in. (45.2 x 22.9 x 32 cm) Wt 58 lb (26 kg)Model 1376N specificationsFree Air Displacement 10.6 cfm (300 L/min)Max. vacuum 1 x 10-4 torrOil capacity 2.5 qt (2.37 L)Tubing needed I.D. 13/16 in.Exhaust and inta
Catalog No. 50-290-8678
Add to Cart
Request a Quote