Learn More
MSC MAPA Professional Heat Resistant Glove, Nitrile, Thermal Acrylic Lined, 250 °F
Catalog No. 502835212
Encompass
The TempDex 710 glove is designed for thermal protection, offering comfort and durability for various industrial applications. It features a thin, anatomically-shaped design for dexterity and excellent abrasion resistance for long-term wear. The glove is free of harmful substances and silicone, preventing defects on materials during handling.
  • Thin, anatomically-shaped design ensures dexterity.
  • Excellent abrasion resistance for long-term wear.
  • Free of harmful substances (DMF free).
  • Silicone-free to prevent traces on metal sheets and glass.
  • Optimized individual packaging, containing 30% recycled material.
  • Dot embossing for better insulation and grip.
  • Suitable for handling hot piping, hot steel after welding, and moulds in the ceramic industry.
  • Rated for heat and fire protection under EN 407 (X1XXXX).
  • Classified for mechanical hazards EN 388 (4111X).
Catalog No. 50-283-5212 Supplier MSC Supplier No. 70202304
The TempDex 710 glove is designed for thermal protection, offering comfort and durability for various industrial applications. It features a thin, anatomically-shaped design for dexterity and excellent abrasion resistance for long-term wear. The glove is free of harmful substances and silicone, preventing defects on materials during handling.
  • Thin, anatomically-shaped design ensures dexterity.
  • Excellent abrasion resistance for long-term wear.
  • Free of harmful substances (DMF free).
  • Silicone-free to prevent traces on metal sheets and glass.
  • Optimized individual packaging, containing 30% recycled material.
  • Dot embossing for better insulation and grip.
  • Suitable for handling hot piping, hot steel after welding, and moulds in the ceramic industry.
  • Rated for heat and fire protection under EN 407 (X1XXXX).
  • Classified for mechanical hazards EN 388 (4111X).