Learn More
Grainger Aquasol Welding Alignment Stick, Water Soluble, 3/8 in x 4 in, 50/Pk.
Catalog No. 502777572
Encompass_Preferred
SoluShim® Water Soluble Alignment Sticks are designed to maintain precise seam spacing for plate, pipe, and flange welding applications. Composed of an EPA-approved water-soluble composite board, these sticks provide a uniform, incompressible alternative to traditional filler rods. They eliminate the need for mechanical grinding or torch cutting during spacer removal, as they dissolve quickly and cleanly when exposed to water or steam, ensuring no residual contamination.
  • Maintains consistent root gaps required by AWS D1.1 – D1.6 and API 1101 codes.
  • Compatible with any metal type.
  • Highly durable, incompressible structure prevents gap shrinkage during tack welding.
  • Environmentally friendly and 100% biodegradable composition.
  • Fast, effortless removal using water or steam.
  • Flexible design allows for easy manual shaping to fit various angles.
Catalog No. 50-277-7572 Supplier Grainger Supplier No. 18C621
SoluShim® Water Soluble Alignment Sticks are designed to maintain precise seam spacing for plate, pipe, and flange welding applications. Composed of an EPA-approved water-soluble composite board, these sticks provide a uniform, incompressible alternative to traditional filler rods. They eliminate the need for mechanical grinding or torch cutting during spacer removal, as they dissolve quickly and cleanly when exposed to water or steam, ensuring no residual contamination.
  • Maintains consistent root gaps required by AWS D1.1 – D1.6 and API 1101 codes.
  • Compatible with any metal type.
  • Highly durable, incompressible structure prevents gap shrinkage during tack welding.
  • Environmentally friendly and 100% biodegradable composition.
  • Fast, effortless removal using water or steam.
  • Flexible design allows for easy manual shaping to fit various angles.