SoluShim alignment sticks are designed to provide consistent seam spacing for plate and pipe welding applications. Constructed from a water-soluble composite board, these adhesive-coated shims are compatible with any metal and help maintain the required root gap during fit-up. The pre-sized, uniform, and flexible sticks stay firmly in place until removed, which is achieved easily by moistening with water. They are 100% biodegradable and leave no harmful trace elements.
- Maintains required root gap during welding
- Highly durable, flexible, and incompressible structure
- Compatible with all metals
- Eliminates costly post-weld grinding
- Conforms to AWS D1.1-D1.6 and API 1101 pipe codes
- Environmentally friendly and 100% biodegradable
- Fast and effortless removal via water dissolution
- Package includes 50 pieces