Learn More
Grainger Aquasol Welding Alignment Stick, Water Soluble, 50/Pk.
Catalog No. 502773698
Encompass_Preferred
SoluShim alignment sticks are designed to provide consistent seam spacing during plate and pipe welding applications. These pre-sized, incompressible spacers maintain the required root gap and prevent shrinkage during tack welding, eliminating the need for metal spacers that require costly grinding. The sticks are compatible with any metal and stay firmly in place throughout the welding process.
  • Maintains root gap during pipe and plate welding
  • Highly durable and incompressible construction
  • Biodegradable and environmentally friendly composition
  • Fast removal via dissolution in water or steam
  • Leaves no harmful residue or post-weld contamination
  • Flexible for manual shaping to any angle
  • Compliant with AWS D1.1 to D1.6 and API 1101 standards
Catalog No. 50-277-3698 Supplier Grainger Supplier No. 18C623
SoluShim alignment sticks are designed to provide consistent seam spacing during plate and pipe welding applications. These pre-sized, incompressible spacers maintain the required root gap and prevent shrinkage during tack welding, eliminating the need for metal spacers that require costly grinding. The sticks are compatible with any metal and stay firmly in place throughout the welding process.
  • Maintains root gap during pipe and plate welding
  • Highly durable and incompressible construction
  • Biodegradable and environmentally friendly composition
  • Fast removal via dissolution in water or steam
  • Leaves no harmful residue or post-weld contamination
  • Flexible for manual shaping to any angle
  • Compliant with AWS D1.1 to D1.6 and API 1101 standards