Learn More
Grainger Aquasol Welding Alignment Stick, Water Soluble, 3/8 in. x 4 in., 50/Pk.
Catalog No. 502777854
Encompass_Preferred
These water soluble alignment sticks provide consistent seam spacing for pipe, sheet, and plate welding applications. Manufactured from an EPA-approved composite board, they offer a durable and incompressible alternative to traditional metal spacers, ensuring precision during fit-up and welding processes.
  • Compatible with any metal type
  • Maintains required root gap throughout welding process
  • Eliminates need for holding rods
  • Prevents root gap shrinkage during tack welding
  • Easily removed by flushing with water or steam
  • Leaves no harmful trace residue
  • Eliminates need for post-weld grinding
  • Highly durable and environmentally friendly
Catalog No. 50-277-7854 Supplier Grainger Supplier No. 18C622
These water soluble alignment sticks provide consistent seam spacing for pipe, sheet, and plate welding applications. Manufactured from an EPA-approved composite board, they offer a durable and incompressible alternative to traditional metal spacers, ensuring precision during fit-up and welding processes.
  • Compatible with any metal type
  • Maintains required root gap throughout welding process
  • Eliminates need for holding rods
  • Prevents root gap shrinkage during tack welding
  • Easily removed by flushing with water or steam
  • Leaves no harmful trace residue
  • Eliminates need for post-weld grinding
  • Highly durable and environmentally friendly