Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Welding Cable 6AWG Neoprene Re

Supplier:  Grainger 19YE21

Encompass_Preferred

Welding Cable 6AWG Neoprene Red 10ft

Catalog No. 50-279-2856


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.