Learn More
MSC Fibre-Metal Welding Safety Plate, Tigerhood Futura, XXL, 5/Pk.
Catalog No. 5000509461
Encompass
The Tigerhood Futura headgear is designed for professionals requiring high levels of protection and visibility in industrial settings. Engineered from lightweight, durable thermoplastics, this welding helmet features an ultra-smooth, reflective silver shell with a streamlined design, suitable for applications including MIG, MAG, TIG, and plasma cutting.
  • Includes an internal safety plate for enhanced protection against impacts
  • Features ADC Plus ADF technology with a 1/1/1/1 DIN optical rating
  • Offers selectable dark state shades 9-13 and a light state of 4
  • Equipped with two sensors and adjustable sensitivity and delay settings
  • Powered by solar energy for efficient, long-term use
  • Includes classic 3-C free-floating headgear for a secure, ergonomic fit
  • Compatible with standard 4 in. x 5 in. front cover lenses and magnifying lenses
Catalog No. 50-005-09461 Supplier MSC Supplier No. 91071118
The Tigerhood Futura headgear is designed for professionals requiring high levels of protection and visibility in industrial settings. Engineered from lightweight, durable thermoplastics, this welding helmet features an ultra-smooth, reflective silver shell with a streamlined design, suitable for applications including MIG, MAG, TIG, and plasma cutting.
  • Includes an internal safety plate for enhanced protection against impacts
  • Features ADC Plus ADF technology with a 1/1/1/1 DIN optical rating
  • Offers selectable dark state shades 9-13 and a light state of 4
  • Equipped with two sensors and adjustable sensitivity and delay settings
  • Powered by solar energy for efficient, long-term use
  • Includes classic 3-C free-floating headgear for a secure, ergonomic fit
  • Compatible with standard 4 in. x 5 in. front cover lenses and magnifying lenses