Learn More
MSC 3M Welding Helmet, Auto-Darkening, PPA, Shade 8 to 12
Catalog No. 502834937
Encompass
This welding helmet offers optimal protection and clear visibility with an auto-darkening lens featuring a shade range of 8 to 12. Constructed from lightweight polyphthalamide (PPA) for resistance to chemicals, heat, and scratches, it also features a versatile lift-front style.
  • Designed for optimal protection and clear visibility.
  • Auto-darkening lens with a shade range of 8 to 12.
  • Constructed from polyphthalamide (PPA), a lightweight and resistant thermoplastic resin.
  • Resistant to chemicals, heat, and scratches.
  • Features a lift-front style.
  • Window dimensions are 5.9 in width and 2.9 in height.
  • Black helmet color.
Catalog No. 50-283-4937 Supplier MSC Supplier No. 16689507
This welding helmet offers optimal protection and clear visibility with an auto-darkening lens featuring a shade range of 8 to 12. Constructed from lightweight polyphthalamide (PPA) for resistance to chemicals, heat, and scratches, it also features a versatile lift-front style.
  • Designed for optimal protection and clear visibility.
  • Auto-darkening lens with a shade range of 8 to 12.
  • Constructed from polyphthalamide (PPA), a lightweight and resistant thermoplastic resin.
  • Resistant to chemicals, heat, and scratches.
  • Features a lift-front style.
  • Window dimensions are 5.9 in width and 2.9 in height.
  • Black helmet color.