Learn More
MSC Lincoln Electric Welding Helmet, Auto-Darkening, Nylon, Shade 5-13, Ratchet Adjustment
Catalog No. 502835204
Encompass
Lincoln Electric welding helmets are designed for high-speed applications. This helmet features an auto-darkening lens and durable nylon material, making it suitable for various welding tasks.
  • Auto-darkening lens for enhanced safety and convenience.
  • Durable nylon material which is breathable, lightweight, and heat resistant.
  • Wide shade range from 5 to 13 for versatility across different welding applications.
  • Ratchet adjustment for a secure and comfortable fit.
  • Dual power source (solar; battery) for reliable operation.
  • Fixed front window style.
  • Features 4 arc sensors for responsive auto-darkening.
Catalog No. 50-283-5204 Supplier MSC Supplier No. 96863063
Lincoln Electric welding helmets are designed for high-speed applications. This helmet features an auto-darkening lens and durable nylon material, making it suitable for various welding tasks.
  • Auto-darkening lens for enhanced safety and convenience.
  • Durable nylon material which is breathable, lightweight, and heat resistant.
  • Wide shade range from 5 to 13 for versatility across different welding applications.
  • Ratchet adjustment for a secure and comfortable fit.
  • Dual power source (solar; battery) for reliable operation.
  • Fixed front window style.
  • Features 4 arc sensors for responsive auto-darkening.