Learn More
MSC WELDING HELMETS
Catalog No. 502834639
Encompass
This welding helmet features digital controls and an auto-darkening lens, suitable for various welding applications. Constructed from black nylon, it offers a fixed front window and a wide range of lens shades. The helmet is solar-powered, lightweight, and heat-resistant.
  • Digital controls for precise settings
  • Auto-darkening lens for enhanced safety
  • Shade range of 4 & 9 to 13
  • Black nylon construction, breathable and heat-resistant
  • Solar-powered operation
  • Fixed front window with 4.0000 in width and 2.0000 in height
  • Field of view 2.5000 in width and 4.0000 in height
Catalog No. 50-283-4639 Supplier MSC Supplier No. 90098203
This welding helmet features digital controls and an auto-darkening lens, suitable for various welding applications. Constructed from black nylon, it offers a fixed front window and a wide range of lens shades. The helmet is solar-powered, lightweight, and heat-resistant.
  • Digital controls for precise settings
  • Auto-darkening lens for enhanced safety
  • Shade range of 4 & 9 to 13
  • Black nylon construction, breathable and heat-resistant
  • Solar-powered operation
  • Fixed front window with 4.0000 in width and 2.0000 in height
  • Field of view 2.5000 in width and 4.0000 in height