Learn More
Grainger Coxreels Welding Hose Reel, 1/4 in I.D., 100 ft, 200 psi, 1 ea
Catalog No. 502767899
Encompass_Preferred
This professional-grade, spring-driven welding hose reel is designed for efficient management of twin-line hoses. Featuring a robust, single-pedestal steel chassis, the unit utilizes a dual axle support system for stability, reduced vibration, and structural integrity. Finished with a durable, chip-resistant powder coat, it includes 100 ft. of 1/4 in. I.D. hose.
  • External 90° full-flow dual-ported brass swivel with Viton® seals allows for side-entry connections.
  • Enclosed, factory-tuned, and removable cartridge-style spring motor.
  • Multi-position non-sparking locking ratchet mechanism secures the hose at desired lengths.
  • CNC robotically spun, ribbed steel discs with rolled edges.
  • Guide arm adjusts for wall, floor, ceiling, or vehicle mounting.
  • Rated for 200 psi max pressure and temperatures of -20°F to 150°F.
Catalog No. 50-276-7899 Supplier Grainger Supplier No. 16X587
This professional-grade, spring-driven welding hose reel is designed for efficient management of twin-line hoses. Featuring a robust, single-pedestal steel chassis, the unit utilizes a dual axle support system for stability, reduced vibration, and structural integrity. Finished with a durable, chip-resistant powder coat, it includes 100 ft. of 1/4 in. I.D. hose.
  • External 90° full-flow dual-ported brass swivel with Viton® seals allows for side-entry connections.
  • Enclosed, factory-tuned, and removable cartridge-style spring motor.
  • Multi-position non-sparking locking ratchet mechanism secures the hose at desired lengths.
  • CNC robotically spun, ribbed steel discs with rolled edges.
  • Guide arm adjusts for wall, floor, ceiling, or vehicle mounting.
  • Rated for 200 psi max pressure and temperatures of -20°F to 150°F.