Learn More
MSC WELDING LENS
Catalog No. 5000677905
Encompass
This 3M cover lens is designed for welding applications. Made from durable nylon and polycarbonate, it features a shade 11 lens for protection and is intended for use with 9100 Series helmets.
  • Black lens color
  • Constructed from nylon and polycarbonate
  • Inside mount type
  • Lens shade 11
  • Compatible with all 9100 Series helmets
  • Package includes 2 lenses
Catalog No. 50-006-77905 Supplier MSC Supplier No. 16988107
This 3M cover lens is designed for welding applications. Made from durable nylon and polycarbonate, it features a shade 11 lens for protection and is intended for use with 9100 Series helmets.
  • Black lens color
  • Constructed from nylon and polycarbonate
  • Inside mount type
  • Lens shade 11
  • Compatible with all 9100 Series helmets
  • Package includes 2 lenses