Learn More
MSC WELDING LENS
Catalog No. 5000676863
Encompass
This welding cover lens is designed for use with 9100 Series welding helmets. It provides essential protection and maintains clear visibility during welding operations.
  • Black lens color
  • Nylon and polycarbonate construction
  • Inside mount type
  • Window dimensions: 7.112 in. height, 10.668 in. width
  • Lens shade 12
  • Designed for 9100 Series welding helmets
  • Manufactured in Czechia
Catalog No. 50-006-76863 Supplier MSC Supplier No. 16988123
This welding cover lens is designed for use with 9100 Series welding helmets. It provides essential protection and maintains clear visibility during welding operations.
  • Black lens color
  • Nylon and polycarbonate construction
  • Inside mount type
  • Window dimensions: 7.112 in. height, 10.668 in. width
  • Lens shade 12
  • Designed for 9100 Series welding helmets
  • Manufactured in Czechia