Learn More
Grainger Welding Table 60x30 In.
Catalog No. 502767928
Encompass_Preferred
The PRO-LINE Welding Table is a 30 inch square work table made of 1/8 inch thick steel with a durable powder coat finish. It features four removable 1/2 inch round bar legs, adjustable leveling feet, and two T-slot tracks for clamping. This sturdy, portable work table is ideal for welding, fabrication, assembly, and other metalworking tasks.
Catalog No. 50-276-7928 Supplier Grainger Supplier No. 16Y918
The PRO-LINE Welding Table is a 30 inch square work table made of 1/8 inch thick steel with a durable powder coat finish. It features four removable 1/2 inch round bar legs, adjustable leveling feet, and two T-slot tracks for clamping. This sturdy, portable work table is ideal for welding, fabrication, assembly, and other metalworking tasks.