Learn More
Grainger Welding Table 72x30 In.
Catalog No. 502767609
Encompass_Preferred
The Pro-Line Welding Table is a 30 inch square work table made of 11 gauge steel with 1/8 inch holes punched 2 inches apart. It has four heavy duty locking casters and can support up to 500 lbs. The table top is 1 inch thick for durability. This rugged portable work table is ideal for welding, fabrication, assembly and other shop projects.
Catalog No. 50-276-7609 Supplier Grainger Supplier No. 16Y919
The Pro-Line Welding Table is a 30 inch square work table made of 11 gauge steel with 1/8 inch holes punched 2 inches apart. It has four heavy duty locking casters and can support up to 500 lbs. The table top is 1 inch thick for durability. This rugged portable work table is ideal for welding, fabrication, assembly and other shop projects.