Learn More
Electron Microscopy Sciences 24/30 mm Well Bar Set
SDP
Catalog No. NC0870439
Encompass_Preferred
We offer Well Bar Sets to make your ordering easier and to offer you a complete set for facilitate your work. Includes: One Well Bar with two wells 24 mm and two wells 30mm (61712-34); Two Large Chucks (61720-36); Two Small Chucks (61720-28); Two Over Chuck Freezing Blocks (61625-01); 8 Dispensing Slides (61727-08).
Catalog No. NC0870439 Supplier Electron Microscopy Sciences Supplier No. 6170404
We offer Well Bar Sets to make your ordering easier and to offer you a complete set for facilitate your work. Includes: One Well Bar with two wells 24 mm and two wells 30mm (61712-34); Two Large Chucks (61720-36); Two Small Chucks (61720-28); Two Over Chuck Freezing Blocks (61625-01); 8 Dispensing Slides (61727-08).