Learn More
Thomson Instrument Company 24 Position Vial Block, for 4mL Vials
SDP
Catalog No. 50996053
Encompass_Preferred
Clear 24-position well block designed for 4 mL vials and smooth handoff between prep steps and automated systems. The uniform layout supports consistent placement, labeling, and scanning while saving deck space. 25/case. Made in the USA
Catalog No. 50-996-053 Supplier Thomson Instrument Company Supplier No. 1212280
Clear 24-position well block designed for 4 mL vials and smooth handoff between prep steps and automated systems. The uniform layout supports consistent placement, labeling, and scanning while saving deck space. 25/case. Made in the USA